Learn More
Cayman Chemical Fluvoxamine (maleate), Purity: ≥95%, CAS: 61718-82-9, 50 mg
SDP
Catalog No. 502968505
Encompass_Preferred
An SSRI; selectively inhibits reuptake of serotonin over norepinephrine reuptake in rat hypothalamic synaptosomes (Kis = 6.2 and 1,100 nM, respectively); reduces immobility time in the forced swim test in mice at 10 mg/kg, an effect that can be blocked by a σ1 receptor antagonist; reduces marble-burying behavior in the marble-burying test in mice at 45 mg/kg
Catalog No. 50-296-8505 Supplier Cayman Chemical Supplier No. 1561750 MG
An SSRI; selectively inhibits reuptake of serotonin over norepinephrine reuptake in rat hypothalamic synaptosomes (Kis = 6.2 and 1,100 nM, respectively); reduces immobility time in the forced swim test in mice at 10 mg/kg, an effect that can be blocked by a σ1 receptor antagonist; reduces marble-burying behavior in the marble-burying test in mice at 45 mg/kg