Learn More
Medchemexpress LLC HY-108690 10mg Medchemexpress, Fluxametamide CAS:928783-29-3 Purity:>98%
SDP
Catalog No. 501968608
Encompass_Preferred
Medchemexpress, HY-108690 10mg Fluxametamide CAS:928783-29-3 Fluxametamide is an insecticide with wide spectrum, acts as an antagonist of GABA- and glutamate-gated chloride channels, with IC50 of 1.95 nM and 225 nM for M. domestica GABACls and GluCls. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8608 Supplier Medchemexpress LLC Supplier No. HY10869010MG
Medchemexpress, HY-108690 10mg Fluxametamide CAS:928783-29-3 Fluxametamide is an insecticide with wide spectrum, acts as an antagonist of GABA- and glutamate-gated chloride channels, with IC50 of 1.95 nM and 225 nM for M. domestica GABACls and GluCls. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.