Learn More
Medchemexpress LLC Fluzoparib 50mg | 1358715-18-0 | 50 MG
SDP
Catalog No. 5000307975
Encompass_Preferred
Fluzoparib is a potent, orally active PARP1 inhibitor supplied for preclinical oncology research, used to study DNA damage repair and tumor responses in BRCA-mutant models.
  • Potent PARP1 inhibitor with low nanomolar enzymatic activity.
  • Orally active compound suitable for in vivo and in vitro studies.
  • Used to study DNA damage repair and BRCA1/2-mutant tumor responses.
  • Provided as a measured solid for precise dosing in preclinical experiments.
  • Molecular formula C22H16F4N6O2 and molecular weight 472.40 to support formulation and dosing calculations.
Catalog No. 50-003-07975 Supplier Medchemexpress LLC Supplier No. HY11477850MG
Fluzoparib is a potent, orally active PARP1 inhibitor supplied for preclinical oncology research, used to study DNA damage repair and tumor responses in BRCA-mutant models.
  • Potent PARP1 inhibitor with low nanomolar enzymatic activity.
  • Orally active compound suitable for in vivo and in vitro studies.
  • Used to study DNA damage repair and BRCA1/2-mutant tumor responses.
  • Provided as a measured solid for precise dosing in preclinical experiments.
  • Molecular formula C22H16F4N6O2 and molecular weight 472.40 to support formulation and dosing calculations.