Learn More
Julabo USA Inc FLW7006 COOLER -20+40C
SDP
Catalog No. 507533965
Encompass_Preferred
FLW7006 water-cooled recirculating cooler with 7.4 kW cooling power at 20 °C; 5.5 kW at 0 °C; 3.1 kW at -10 °C, 1.3 kW at -20 °C for operation from -20…+40 °C. Proportional cooling control. Filling volume 39…47 L with sight glass. Centrifugal pump to 60 L/min with maximum pressure of 87 psi (6 bar). Integrated internal by-pass for pressure adjustment with analog pressure gauge. G 1 1/4" inlet/outlet connections with 1" hose barbs. RS232 and Stakei outlets. Dimensions 30.7x33.5x58.3 inches (WxLxH); 485 pounds. 230V/3PPE/60Hz
Catalog No. 50-753-3965 Supplier Julabo USA Inc Supplier No. 9676070.16
Add to Cart
Add to Cart
Request a Quote
FLW7006 water-cooled recirculating cooler with 7.4 kW cooling power at 20 °C; 5.5 kW at 0 °C; 3.1 kW at -10 °C, 1.3 kW at -20 °C for operation from -20…+40 °C. Proportional cooling control. Filling volume 39…47 L with sight glass. Centrifugal pump to 60 L/min with maximum pressure of 87 psi (6 bar). Integrated internal by-pass for pressure adjustment with analog pressure gauge. G 1 1/4" inlet/outlet connections with 1" hose barbs. RS232 and Stakei outlets. Dimensions 30.7x33.5x58.3 inches (WxLxH); 485 pounds. 230V/3PPE/60Hz