Learn More
Julabo USA Inc FLW7006 CHILLE -20+40C, 7.4kW cooling, 6 bar pump, 208-230V/3PPE/60Hz
SDP
Catalog No. 507533962
Encompass_Preferred
FL7006 water-cooled recirculating cooler with 7.4 kW cooling power at 20 C; 5.1 kW at 0 C; 3 kW at -10 C, 1.55 kW at -20 C for operation from -20 +40 C. Proportional cooling control. Filling volume 39 47 L with sight glass. Centrifugal pump to 60 L/min with maximum pressure of 87 psi (6 bar). Integrated internal by-pass for pressure adjustment with analog pressure gauge. G 1 1/4" inlet/outlet connections with 1" hose barbs. RS232 and Stakei outlets. Dimensions 30.7x33.5x58.3 inches (WxLxH); 556 pounds. 230V/3PPE/60Hz
Catalog No. 50-753-3962 Supplier Julabo USA Inc Supplier No. 9666070.16
Add to Cart
Add to Cart
Request a Quote
FL7006 water-cooled recirculating cooler with 7.4 kW cooling power at 20 C; 5.1 kW at 0 C; 3 kW at -10 C, 1.55 kW at -20 C for operation from -20 +40 C. Proportional cooling control. Filling volume 39 47 L with sight glass. Centrifugal pump to 60 L/min with maximum pressure of 87 psi (6 bar). Integrated internal by-pass for pressure adjustment with analog pressure gauge. G 1 1/4" inlet/outlet connections with 1" hose barbs. RS232 and Stakei outlets. Dimensions 30.7x33.5x58.3 inches (WxLxH); 556 pounds. 230V/3PPE/60Hz