Learn More
Microsolv Technology Corporation Flx Peek Tub.005 Id20Mm Red
Catalog No. 5001451651
Encompass_Preferred
Tubing, PEEK, Coiled, 0.005 ID, 1/16 OD. 945mm L, General Grade, Red, with Tubing Connectors. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51651 Supplier Microsolv Technology Corporation Supplier No. 49306502R
Tubing, PEEK, Coiled, 0.005 ID, 1/16 OD. 945mm L, General Grade, Red, with Tubing Connectors. MicroSolv Brand. 1/EA.