Learn More
Microsolv Technology Corporation Flx Peek Tub.020 Id20Mm Ora
Catalog No. 5001451653
Encompass_Preferred
Tubing, PEEK, Coiled, 0.020 ID, 1/16 OD. 945mm L, General Grade, Orange, with Tubing Connectors. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51653 Supplier Microsolv Technology Corporation Supplier No. 49306508E
Tubing, PEEK, Coiled, 0.020 ID, 1/16 OD. 945mm L, General Grade, Orange, with Tubing Connectors. MicroSolv Brand. 1/EA.