Learn More
MSC Brown and Sharpe Flexible Steel Rule, 12 in. Length, 1/2 in. Width, Satin-Chrome Finish, 1 ea.
Catalog No. 5000510212
Encompass
This flexible steel rule is a precision measuring tool manufactured from high-quality hardened and tempered steel. Designed for durability and clarity, it features a satin-chrome finish that reduces glare, making it suitable for use in varied industrial environments.
  • Hardened and tempered steel construction for long-lasting use
  • Satin-chrome finish provides excellent readability
  • Includes 32nds and 64ths imperial graduations
  • Includes mm and 1/2 mm metric graduations
  • Measures 12 in. long by 1/2 in. wide
  • Thickness of 0.015 in.
Catalog No. 50-005-10212 Supplier MSC Supplier No. 99235962
This flexible steel rule is a precision measuring tool manufactured from high-quality hardened and tempered steel. Designed for durability and clarity, it features a satin-chrome finish that reduces glare, making it suitable for use in varied industrial environments.
  • Hardened and tempered steel construction for long-lasting use
  • Satin-chrome finish provides excellent readability
  • Includes 32nds and 64ths imperial graduations
  • Includes mm and 1/2 mm metric graduations
  • Measures 12 in. long by 1/2 in. wide
  • Thickness of 0.015 in.