Learn More
Grainger J.T. Eaton Insect Trap, Sticky Tape, 14 ft, 1 ea
Catalog No. 502785636
Encompass_Preferred
This fly ribbon on a roll provides effective, chemical-free control for flies and other flying insects. The adhesive trap is designed for both indoor and outdoor use, offering a non-toxic solution that is safe for use around pets and people. The ribbon is sticky on both sides to maximize surface area and features a bright white color to attract pests. With 14 ft. of ribbon per roll, users can pull down the desired length and snip off used portions as needed.
  • Sticky on both sides to double available capture area
  • Non-toxic and free of vapors, poisons, and odors
  • Durable adhesive remains effective in direct sunlight and will not tear
  • Adjustable 14 ft. length with included mounting hardware
  • Safe for use in kitchens, trash areas, barns, stables, and warehouses
  • Ready-to-use design with easy disposal
Catalog No. 50-278-5636 Supplier Grainger Supplier No. 196Y99
This fly ribbon on a roll provides effective, chemical-free control for flies and other flying insects. The adhesive trap is designed for both indoor and outdoor use, offering a non-toxic solution that is safe for use around pets and people. The ribbon is sticky on both sides to maximize surface area and features a bright white color to attract pests. With 14 ft. of ribbon per roll, users can pull down the desired length and snip off used portions as needed.
  • Sticky on both sides to double available capture area
  • Non-toxic and free of vapors, poisons, and odors
  • Durable adhesive remains effective in direct sunlight and will not tear
  • Adjustable 14 ft. length with included mounting hardware
  • Safe for use in kitchens, trash areas, barns, stables, and warehouses
  • Ready-to-use design with easy disposal