Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC J.T. Eaton Window Trap, Targets Flies, 5 Traps

Catalog No. 502869649
Encompass

This window trap is designed to effectively capture flies. It utilizes a clear to pale yellow tacky solid adhesive, insoluble in water, and exhibits a characteristic petroleum odor.

  • Trap type: Window trap
  • Chemical family: Hot melt adhesive/thermoplastic
  • Melting point: 325° F
  • Specific gravity (H2O = 1): 0.93
  • Country of origin: China

Catalog No. 50-286-9649 Supplier MSC Supplier No. 51351823
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This window trap is designed to effectively capture flies. It utilizes a clear to pale yellow tacky solid adhesive, insoluble in water, and exhibits a characteristic petroleum odor.

  • Trap type: Window trap
  • Chemical family: Hot melt adhesive/thermoplastic
  • Melting point: 325° F
  • Specific gravity (H2O = 1): 0.93
  • Country of origin: China

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.