Learn More
ARBOR SCIENTIFIC FLYING PIG
SDP
Catalog No. NC0369506
Encompass
Add some excitement to your lessons on circular motion with the Flying Pig. Suspended from the ceiling, his wings flap and carry him in a regular circle. With the string, this motion gives students a conical pendulum.
Catalog No. NC0369506 Supplier ARBOR SCIENTIFIC Supplier No. P42165
Add some excitement to your lessons on circular motion with the Flying Pig. Suspended from the ceiling, his wings flap and carry him in a regular circle. With the string, this motion gives students a conical pendulum.