Learn More
Genesee Scientific Corporation FLYSTUFF FLYPAD,8.1 X 11.6CM
SDP
Catalog No. NC1945895
Encompass
CO2 Anesthetizing Apparatus1 Flypad/UnitPrecision routed flow maze ensures optimal, consistent diffusion of CO2High-quality construction maintains a flat, warp free surface made of UHMW porous polyethyleneIncludes nickel-plated brass threaded hose barb input compatable with 1/8" ID (3mm) tubingDimensions: 8.1 x 11.6cm
Catalog No. NC1945895 Supplier Genesee Scientific Corporation Supplier No. 59114
Add to Cart
Add to Cart
CO2 Anesthetizing Apparatus1 Flypad/UnitPrecision routed flow maze ensures optimal, consistent diffusion of CO2High-quality construction maintains a flat, warp free surface made of UHMW porous polyethyleneIncludes nickel-plated brass threaded hose barb input compatable with 1/8" ID (3mm) tubingDimensions: 8.1 x 11.6cm