Learn More
CELLTREAT Scientific Products FlyTreat™ Drosophila Bottle, 8oz, Round, Polypropylene, Non-sterile, Case Quantity 125
SDP
Catalog No. 5000670031
Encompass_Preferred
Support larger Drosophila populations with 6oz and 8oz bottles
Polypropylene material helps mitigate humidity loss compared to other vessels
Bottles provide more room for flies to live and move freely, reducing stress and fostering a healthier environment
Anesthetizing and sorting tasks become easier with more space
Catalog No. 50-006-70031 Supplier CELLTREAT Scientific Products Supplier No. 229956
Support larger Drosophila populations with 6oz and 8oz bottles
Polypropylene material helps mitigate humidity loss compared to other vessels
Bottles provide more room for flies to live and move freely, reducing stress and fostering a healthier environment
Anesthetizing and sorting tasks become easier with more space