Learn More
CELLTREAT Scientific Products FlyTreat™ Drosophila Vial Plug, Polyester, Bottle, Non-sterile, Case Quantity 250
SDP
Catalog No. 5000670040
Encompass_Preferred
Breathable polyester plugs minimize mite contaminationand fly escape
Autoclavable and reusable
Catalog No. 50-006-70040 Supplier CELLTREAT Scientific Products Supplier No. 229909
Breathable polyester plugs minimize mite contaminationand fly escape
Autoclavable and reusable