Learn More
Aviva Systems Biology Corp FLYWCH1 Antibody - middle region (ARP60443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLYWCH1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FLYWCH1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-047