Learn More
Selleck Chemical LLC FM-381 2mg 2226521-65-7
SDP
Supplier:  Selleck Chemical LLC S85412MG
Encompass_Preferred
FM-381 is a JAK3 specific reversible covalent inhibitor with IC50 of 127 pM for JAK3 and demonstrates 400-, 2,700- and 3,600-fold selectivity over JAK1, JAK2, and TYK2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8246