Learn More
Selleck Chemical LLC FM-381
SDP
Catalog No. 5000001483
Encompass_Preferred
FM-381 is a JAK3 specific reversible covalent inhibitor with IC50 of 127 pM for JAK3 and demonstrates 400- 2 700- and 3 600-fold selectivity over JAK1 JAK2 and TYK2 respectively
Catalog No. 50-000-01483 Supplier Selleck Chemical LLC Supplier No. S85415MG
FM-381 is a JAK3 specific reversible covalent inhibitor with IC50 of 127 pM for JAK3 and demonstrates 400- 2 700- and 3 600-fold selectivity over JAK1 JAK2 and TYK2 respectively