Learn More
TARGETMOL CHEMICALS INC FM-381 5MG
SDP
Catalog No. 502877336
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. FM381 is a potent covalent reversible inhibitor of JAK3. FM-381 has an IC50 of 127 pM for JAK3, with 410, 2700 and 3600-fold selectivity over JAK1, JAK2 and TYK2, respectively. Purity 97.71%
Catalog No. 50-287-7336 Supplier TARGETMOL CHEMICALS INC Supplier No. T50915MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. FM381 is a potent covalent reversible inhibitor of JAK3. FM-381 has an IC50 of 127 pM for JAK3, with 410, 2700 and 3600-fold selectivity over JAK1, JAK2 and TYK2, respectively. Purity 97.71%