Learn More
Med Vet International Dynarex Sterile Dynalevin Waterproof Adhesive Bordered Foam, 4" X 4", 120/Case
Catalog No. 502831115
Encompass_Preferred
A sterile, foam with waterproof outer layer and adhesive border used to maintain a moist healing environment, while helping keep contaminants out. Made of absorbent foam with waterproof outer layer and adhesive border that adheres easily to skin, helping to keep contaminants out to accelerate healing. Medium absorbency dressing used to provide an optimal, moist wound healing environment for exuding wounds. Primary or secondary dressing used to manage chronic and acute wounds at stages 2, 3, and 4.
Catalog No. 50-283-1115 Supplier Med Vet International Supplier No. 3021
A sterile, foam with waterproof outer layer and adhesive border used to maintain a moist healing environment, while helping keep contaminants out. Made of absorbent foam with waterproof outer layer and adhesive border that adheres easily to skin, helping to keep contaminants out to accelerate healing. Medium absorbency dressing used to provide an optimal, moist wound healing environment for exuding wounds. Primary or secondary dressing used to manage chronic and acute wounds at stages 2, 3, and 4.