Learn More
Cayman Chemical α-Arbutin, Purity: ≥98%, CAS: 84380-01-8, 50 g
SDP
Catalog No. 502913385
Encompass_Preferred
A glycosylated hydroquinone with diverse biological activities; inhibits tyrosinase in a cell-free assay (IC50 = 0.48 mM for the mouse enzyme); inhibits production of melanin in HMV-II cells at 0.5 mM; prevents rotenone-induced increases in ROS in SH-SY5Y cells at 100 µM; decreases locomotor deficits in a parkin-null transgenic Drosophila model of Parkinson's disease; reduces postprandial plasma glucose levels in mice at 30 mM; has been used in the synthesis of α-glucosidase inhibitors
Catalog No. 50-291-3385 Supplier Cayman Chemical Supplier No. 3538650 G
A glycosylated hydroquinone with diverse biological activities; inhibits tyrosinase in a cell-free assay (IC50 = 0.48 mM for the mouse enzyme); inhibits production of melanin in HMV-II cells at 0.5 mM; prevents rotenone-induced increases in ROS in SH-SY5Y cells at 100 µM; decreases locomotor deficits in a parkin-null transgenic Drosophila model of Parkinson's disease; reduces postprandial plasma glucose levels in mice at 30 mM; has been used in the synthesis of α-glucosidase inhibitors