Learn More
Medchemexpress LLC HY-103466 1mg , FM4-64 CAS:162112-35-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034661MG
Encompass_Preferred
HY-103466 1mg FM4-64 CAS: 162112-35-8 FM4-64, a membrane-selective red fluorescent dye, belongs to the FM family of styrylpyridinium dyes. FM4-64 is widely used to study endocytosis and exocytosis, vesicle trafficking and organelle organization in living animal. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1256