Learn More
Medchemexpress LLC FMAU | 1G
SDP
Catalog No. 5000226965
Encompass_Preferred
FMAU is a thymidine analogue Analogs of this series have insertional activity towards replicated DNA They can be used to label cells and track DNA synthesis[1]
Catalog No. 50-002-26965 Supplier Medchemexpress LLC Supplier No. HY1142471G
FMAU is a thymidine analogue Analogs of this series have insertional activity towards replicated DNA They can be used to label cells and track DNA synthesis[1]