Learn More
Cayman Chemical FMF-04-159-2 10mg
SDP
Catalog No. 502978468
Encompass_Preferred
A TAIRE family kinase inhibitor; binds to Cdk14 and Cdk16 (IC50s = 88 and 10.1 nM, respectively); selectively inhibits Cdk14, Cdk16, Cdk17, and Cdk18 over a panel of 16 additional kinases at 1 µM but does inhibit Cdk2 and Cdk10 by greater than 70% at 1 µM
Catalog No. 50-297-8468 Supplier Cayman Chemical Supplier No. 3998910 MG
A TAIRE family kinase inhibitor; binds to Cdk14 and Cdk16 (IC50s = 88 and 10.1 nM, respectively); selectively inhibits Cdk14, Cdk16, Cdk17, and Cdk18 over a panel of 16 additional kinases at 1 µM but does inhibit Cdk2 and Cdk10 by greater than 70% at 1 µM