Learn More
Medchemexpress LLC FMF-04-159-2 | 2364489-81-4 | 99.5% | 1 ML
SDP
Catalog No. 5000342429
Encompass_Preferred
FMF-04-159-2 is a potent covalent inhibitor targeting both CDK14 and CDK2, exhibiting IC50s of 39.6 nM and 256 nM respectively in NanoBRET assays. This compound effectively inhibits HCT116 proliferation with an IC50 of 1,144±190 nM.
  • Covalent CDK14 and CDK2 inhibitor
  • Potent inhibition of HCT116 proliferation
  • High solubility in DMSO (≥ 100 mg/mL)
  • Comprehensive documentation, including Data Sheet, COA, and SDS, is available
Catalog No. 50-003-42429 Supplier Medchemexpress LLC Supplier No. HY12710410MM/1ML
FMF-04-159-2 is a potent covalent inhibitor targeting both CDK14 and CDK2, exhibiting IC50s of 39.6 nM and 256 nM respectively in NanoBRET assays. This compound effectively inhibits HCT116 proliferation with an IC50 of 1,144±190 nM.
  • Covalent CDK14 and CDK2 inhibitor
  • Potent inhibition of HCT116 proliferation
  • High solubility in DMSO (≥ 100 mg/mL)
  • Comprehensive documentation, including Data Sheet, COA, and SDS, is available