Learn More
Medchemexpress LLC HY-127104 5mg Medchemexpress, FMF-04-159-2 CAS:2364489-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1271045MG
Encompass_Preferred
Medchemexpress, HY-127104 5mg FMF-04-159-2 CAS:2364489-81-4 FMF-04-159-2 is a covalent CDK14 inhibitor. FMF-04-159-2 inhibits CDK14 and CDK2 with IC50s of 39.6 nM and 256 nM in NanoBRET assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9738