Learn More
Medchemexpress LLC FMF-04-159-R | 2741262-15-5 | 99.8% | 685.02 | 5 MG
SDP
Catalog No. 5000392690
Encompass_Preferred
FMF-04-159-R is a cyclin-dependent kinase CDK14 and CDK16 inhibitor with IC50 values of 5.9 and 139.1 nM. This compound is suitable for research applications.
  • Acts as a CDK14 and CDK16 inhibitor.
  • Demonstrates potent inhibitory activity with low nanomolar IC50 values.
  • Ideal for research purposes.
Catalog No. 50-003-92690 Supplier Medchemexpress LLC Supplier No. HY1631705MG
FMF-04-159-R is a cyclin-dependent kinase CDK14 and CDK16 inhibitor with IC50 values of 5.9 and 139.1 nM. This compound is suitable for research applications.
  • Acts as a CDK14 and CDK16 inhibitor.
  • Demonstrates potent inhibitory activity with low nanomolar IC50 values.
  • Ideal for research purposes.