Learn More
Medchemexpress LLC FMF-06-098-1 | 2769753-07-1 | 98.77% | 1073.76 | 50 MG
SDP
Catalog No. 5000360619
Encompass_Preferred
FMF-06-098-1 is a multi-target kinase PROTAC degrader, designed to target the degradation of various kinases.
  • Targets the degradation of AAK1, AΒL2, AURKA, AURKB, BUBIB, CDC7, CDK1, CDK12, CDK13, CDK2, CDK4, CDk6, CDK7, CDK9, CHEK1, CSNKID, EPHA1, PER, FGFR1, GAK, IRAK4, ITK, LIMK2, MAP4K2, MAP4K3, MAPK6, MAPK7, MARK4, MELK, PKN3, PLK4, PRKAA1, PTK2, PTK6, RPS6KA4, S1K2, STK35, TNK2, UHMK1, ULK1, and WEE1.
  • Composed of a target protein ligand, a VHL ligand, and a linker.
  • Intended for research use only.
Catalog No. 50-003-60619 Supplier Medchemexpress LLC Supplier No. HY14806550MG
FMF-06-098-1 is a multi-target kinase PROTAC degrader, designed to target the degradation of various kinases.
  • Targets the degradation of AAK1, AΒL2, AURKA, AURKB, BUBIB, CDC7, CDK1, CDK12, CDK13, CDK2, CDK4, CDk6, CDK7, CDK9, CHEK1, CSNKID, EPHA1, PER, FGFR1, GAK, IRAK4, ITK, LIMK2, MAP4K2, MAP4K3, MAPK6, MAPK7, MARK4, MELK, PKN3, PLK4, PRKAA1, PTK2, PTK6, RPS6KA4, S1K2, STK35, TNK2, UHMK1, ULK1, and WEE1.
  • Composed of a target protein ligand, a VHL ligand, and a linker.
  • Intended for research use only.