Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmf-06-098-1 | 2769753-07-1 | 98.8% | 1073.76 | 25 MG
SDP

Catalog No. 5000363791
Encompass_Preferred

FMF-06-098-1 is a multi-target kinase PROTAC degrader that targets degradation kinases such as AAK1, AΒL2, AURKA, AURKB, BUBIB, CDC7, CDK1, CDK12, CDK13, CDK2, CDK4, CDk6, CDK7, CDK9, CHEK1, CSNKID, EPHA1, PER, FGFR1, GAK, IRAK4, ITK, LIMK2, MAP4K2, MAP4K3, MAPK6, MAPK7, MARK4, MELK, PKN3, PLK4, PRKAA1, PTK2, PTK6, RPS6KA4, S1K2, STK35, TNK2, UHMK1, ULK1, and WEE1. The compound is composed of a target protein ligand, a VHL ligand, and a linker.

  • Store powder at -20°C for 3 years, or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.

Catalog No. 50-003-63791 Supplier Medchemexpress LLC Supplier No. 25MGHY148065
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FMF-06-098-1 is a multi-target kinase PROTAC degrader that targets degradation kinases such as AAK1, AΒL2, AURKA, AURKB, BUBIB, CDC7, CDK1, CDK12, CDK13, CDK2, CDK4, CDk6, CDK7, CDK9, CHEK1, CSNKID, EPHA1, PER, FGFR1, GAK, IRAK4, ITK, LIMK2, MAP4K2, MAP4K3, MAPK6, MAPK7, MARK4, MELK, PKN3, PLK4, PRKAA1, PTK2, PTK6, RPS6KA4, S1K2, STK35, TNK2, UHMK1, ULK1, and WEE1. The compound is composed of a target protein ligand, a VHL ligand, and a linker.

  • Store powder at -20°C for 3 years, or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.