Description
- Three channel fluorescence (DAPI, FITC, TRITC)
- Adjustable pillar height
- Viewing up to 10 cell stacks or large vessels
Specifications
Specifications
| Voltage | 100/240 V |
| Magnification Power | 4X, 10X, 20X, 40X |
| Objective Types | LWD, Fluorescence, Brightfield, Color Brightfield, Phase Contrast |
| Eyepiece | 10X/22 mm |
| Lighting Type | LED |
| Head Style | Trinocular, 45° Inclined |
| Camera Type | CMOS/CCD |
| Display Type | None |
| Product Type | Inverted Phase-contrast Fluorescent Microscope |