Learn More
Medchemexpress LLC HY-52101A 2mg , FMK CAS:821794-92-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY52101A2MG
Encompass_Preferred
HY-52101A 2mg FMK CAS: 821794-92-7 FMK is a an irreversible RSK2 kinase inhibitor, that covalently modifies the C-terminal kinase domain of RSK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1091