Learn More
TARGETMOL CHEMICALS INC FMK 5MG
SDP
Catalog No. 502578798
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FMK is an irreversible inhibitor of RSK2 kinase. purity: 98%
Catalog No. 50-257-8798 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ03105MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FMK is an irreversible inhibitor of RSK2 kinase. purity: 98%