Learn More
Medchemexpress LLC HY-100522 1mg Medchemexpress, FMK 9a CAS:1955550-51-2 Purity:>98%
SDP
Catalog No. 501931867
Encompass_Preferred
Medchemexpress, HY-100522 1mg FMK 9a CAS:1955550-51-2 FMK 9a is an autophagin-1 inhibitor with IC 50 values of 80 and 73 μM in FRET and LRA assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1867 Supplier Medchemexpress LLC Supplier No. HY1005221MG
Medchemexpress, HY-100522 1mg FMK 9a CAS:1955550-51-2 FMK 9a is an autophagin-1 inhibitor with IC 50 values of 80 and 73 μM in FRET and LRA assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.