Learn More
TARGETMOL CHEMICALS INC FMK 9A 5MG
SDP
Catalog No. 502581232
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FMK 9a is an irreversible inhibitor of ATG4B with IC50 values of 80 and 73 nM in the TR-FRET and cellular-based LRA assays. purity: 98%
Catalog No. 50-258-1232 Supplier TARGETMOL CHEMICALS INC Supplier No. T153035MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FMK 9a is an irreversible inhibitor of ATG4B with IC50 values of 80 and 73 nM in the TR-FRET and cellular-based LRA assays. purity: 98%