Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Female Luer to Threaded Adapter, 1/4-28 UNF; 25/PK
Catalog No. NC0591679
Encompass
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
Catalog No. NC0591679 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550866
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.