Learn More
Bachem For-Met-Leu-pNA
SDP
Supplier:  Bachem 4027984.0250
Encompass_Preferred
For-Met-Leu-pNA, 111150-07-3, C18H26N4O5S, Mr 410.49, 250mg. fML-pNA, chromogenic substrate for a continuous spectrophotometric assay for peptide deformylase.
Catalog No. 50-194-7067