Learn More
Biotium Forget-Me-Not™ EvaGreen qPCR Master Mix (2-Color Tracking) (500 reactions)
Catalog No. 501964555
Encompass_Preferred
Forget-Me-Not™ EvaGreen qPCR Master Mix is a hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ features a unique combination of a master mix containing a low concentration of blue dye, plus a DNA template buffer containing a higher concentration of blue dye. This 2-color tracking system allows you to see at a glance whether you forgot to add master mix or template to any of your reactions, so you can catch pipetting mistakes and avoid wasting time, reagents, and your precious DNA samples. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols. This product has sufficient reagent for 500 PCR reactions.
Catalog No. 50-196-4555 Supplier Biotium Supplier No. 310411
Forget-Me-Not™ EvaGreen qPCR Master Mix is a hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ features a unique combination of a master mix containing a low concentration of blue dye, plus a DNA template buffer containing a higher concentration of blue dye. This 2-color tracking system allows you to see at a glance whether you forgot to add master mix or template to any of your reactions, so you can catch pipetting mistakes and avoid wasting time, reagents, and your precious DNA samples. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols. This product has sufficient reagent for 500 PCR reactions.