Learn More
Biotium Forget-Me-Not™ EvaGreen qPCR Master Mix (2-Color Tracking) with ROX (2000 reactions)
Supplier:  Biotium 3104220ML
Encompass_Preferred
Forget-Me-Not™ EvaGreen qPCR Master Mix is a hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ features a unique combination of a master mix containing a low concentration of blue dye, plus a DNA template buffer containing a higher concentration of blue dye. This 2-color tracking system allows you to see at a glance whether you forgot to add master mix or template to any of your reactions, so you can catch pipetting mistakes and avoid wasting time, reagents, and your precious DNA samples. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols and ROX reference dye for use with any PCR instrument. This product has sufficient reagent for 2000 PCR reactions.
Catalog No. 50-196-4558