Learn More
Biotium Forget-Me-Not™ EvaGreen qPCR Master Mix (High ROX) (100 reactions)
Catalog No. 501964562
Encompass_Preferred
Forget-Me-Not™ EvaGreen qPCR Master Mix is a high performance, hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ master mix has a light blue master mix color which helps prevent pipetting mistakes, saving time and reagents. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols and High ROX reference dye. This trial size has sufficient reagent for 100 PCR reactions.
Catalog No. 50-196-4562 Supplier Biotium Supplier No. 310461ML
Forget-Me-Not™ EvaGreen qPCR Master Mix is a high performance, hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ master mix has a light blue master mix color which helps prevent pipetting mistakes, saving time and reagents. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols and High ROX reference dye. This trial size has sufficient reagent for 100 PCR reactions.