Learn More
Biotium Forget-Me-Not™ EvaGreen qPCR Master Mix (Low ROX) (500 reactions)
Catalog No. 501964561
Encompass_Preferred
Forget-Me-Not™ EvaGreen qPCR Master Mix is a high performance, hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ master mix has a light blue master mix color which helps prevent pipetting mistakes, saving time and reagents. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols and Low ROX reference dye. This product has sufficient reagent for 500 PCR reactions.
Catalog No. 50-196-4561 Supplier Biotium Supplier No. 310455ML
Forget-Me-Not™ EvaGreen qPCR Master Mix is a high performance, hot-start EvaGreen dye-based master mix for use in real time PCR applications and DNA melt curve analysis. Forget-Me-Not™ master mix has a light blue master mix color which helps prevent pipetting mistakes, saving time and reagents. The master mix features EvaGreen Dye which is suitable for HRM analysis and is safer, more environmentally friendly, and more sensitive than SYBR Green. The kit also features Cheetah™ Hotstart Taq for fast cycling protocols and Low ROX reference dye. This product has sufficient reagent for 500 PCR reactions.