Learn More
Biotium Forget-Me-Not™ Universal Probe Master Mix (500 reactions)
Catalog No. 501964559
Encompass_Preferred
2X Forget-Me-Not™ Universal qPCR Probe Master Mix is a high-performance product for fluorescent probe-based PCR applications, including quantitation and SNP genotyping. This kit is suitable for all fluorescent probe-based technologies, including hydrolysis probes (such as TaqMan and dual-labeled BHQ probes) and displacement probes (like Molecular Beacons). Forget-Me-Not™ Universal Probe Master Mix shows excellent concordance of results in singleplex and multiplex reactions, has broad instrument compatibility, and can be used in both standard and fast protocols. This kit does not contain ROX reference dye. This product has sufficient reagent for 500 PCR reactions.
Catalog No. 50-196-4559 Supplier Biotium Supplier No. 310431
2X Forget-Me-Not™ Universal qPCR Probe Master Mix is a high-performance product for fluorescent probe-based PCR applications, including quantitation and SNP genotyping. This kit is suitable for all fluorescent probe-based technologies, including hydrolysis probes (such as TaqMan and dual-labeled BHQ probes) and displacement probes (like Molecular Beacons). Forget-Me-Not™ Universal Probe Master Mix shows excellent concordance of results in singleplex and multiplex reactions, has broad instrument compatibility, and can be used in both standard and fast protocols. This kit does not contain ROX reference dye. This product has sufficient reagent for 500 PCR reactions.