Learn More
Aviva Systems Biology Corp FMO3 Antibody - N-terminal region (ARP44433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FMO3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FMO3. Alternate names: FMOII, MGC34400, dJ127D3.1, TMAUApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-144