Learn More
Aviva Systems Biology Corp Fmo3 Antibody - middle region (ARP44435_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44435_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fmo3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW111792ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-145