Learn More
Cayman Chemical Fmoc-1-Methyl-L-histidine, Purity: ≥98%, CAS: 202920-22-7, 250 mg
SDP
Catalog No. 502912477
Encompass_Preferred
An amino acid-containing building block; has been used in the production of ε-nitrogen-ligated copper centers in copper nitrite reductases
Catalog No. 50-291-2477 Supplier Cayman Chemical Supplier No. 36312250 MG
An amino acid-containing building block; has been used in the production of ε-nitrogen-ligated copper centers in copper nitrite reductases