Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-10-Adc-OH | 10 g | CAS 143688-82-8 | MDL MFCD00270352

Catalog No. 502349445
Encompass

Fmoc-10-Adc-OH is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 143688-82-8
- MDL: MFCD00270352
- InChIKey: VRQUPGNWTJGCBX-UHFFFAOYSA-N
- Molecular Weight: 409.526
- Molecular Formula: C25H31NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 10-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
- SMILES: O=C(CCCCCCCCCNC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)O

Catalog No. 50-234-9445 Supplier STA Pharmaceutical Supplier No. STAAA1026610GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-10-Adc-OH is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 143688-82-8
- MDL: MFCD00270352
- InChIKey: VRQUPGNWTJGCBX-UHFFFAOYSA-N
- Molecular Weight: 409.526
- Molecular Formula: C25H31NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 10-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
- SMILES: O=C(CCCCCCCCCNC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.