Learn More
Medchemexpress LLC Fmoc-11-aminoundecan 25g
SDP
Catalog No. 5000204968
Encompass_Preferred
Fmoc-11-aminoundecanoic acid can be used as a PROTAC linker in the synthesis of PROTACs and other conjugation applications Fmoc-11-aminoundecanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond
Catalog No. 50-002-04968 Supplier Medchemexpress LLC Supplier No. 25GHYW097110
Fmoc-11-aminoundecanoic acid can be used as a PROTAC linker in the synthesis of PROTACs and other conjugation applications Fmoc-11-aminoundecanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond