Learn More
Medchemexpress LLC Fmoc-12-aminododecan 1g
SDP
Catalog No. 5000204598
Encompass_Preferred
Fmoc-12-aminododecanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The compound can be used as a PROTAC linker in the synthesis of PROTACs The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond
Catalog No. 50-002-04598 Supplier Medchemexpress LLC Supplier No. HYW0971281G
Fmoc-12-aminododecanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The compound can be used as a PROTAC linker in the synthesis of PROTACs The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond