Learn More
STA PHARMACEUTICAL US LLC Fmoc-1,6-Diaminohexane HCl | 5 g | CAS 945923-91-1 | MDL MFCD00830743
Supplier: STA PHARMACEUTICAL US LLC STAAA115155GM
Fmoc-1,6-Diaminohexane HCl is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 945923-91-1
- MDL: MFCD00830743
- InChIKey: SKDQIWLYPJOUMY-UHFFFAOYSA-N
- Molecular Weight: 374.909
- Molecular Formula: C21H27ClN2O2
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (6-aminohexyl)carbamate hydrochloride
- SMILES: O=C(NCCCCCCN)OCC1C2=CC=CC=C2C3=CC=CC=C31.Cl
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.