Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-™-2-(7-Octenyl)Ala-Oh

Catalog No. F65621G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F65621G 100 g
F65625G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. F65621G Supplier Sigma Aldrich Supplier No. F65621G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Olefinic alpha-methyl amino acid for peptide stapling. Upon incorporation of this amino acid into a peptide, along with another of the same or derivative with a different length of the olefinic side chain, the two can be 'stapled' via a ring closing metathesis reaction with Grubb's catalyst (product # 579726). The resulting stapled peptide macrocycle has been shown to stabilize the alpha-helical structure of peptides, which can lead to favorable biological characteristics such as increased proteolytic stability and cellular uptake.

Specifications

CAS 59-30-3
Melting Point > 285°C
Molecular Formula C19H19N7O6
Linear Formula C19H19N7O6
MDL Number MFCD00079305
Quantity 100 g
Molecular Weight (g/mol) 441.4 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.