Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-hPhe(2,5-diF)-OH | 10 g | CAS 1260601-14-6 | MDL MFCD07372306

Catalog No. 502352007
Encompass

Fmoc-D-hPhe(2,5-diF)-OH is a Amino Acid reagent (Subcategory: Homo-natural AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1260601-14-6
- MDL: MFCD07372306
- InChIKey: CMOCQWVOWRXWCO-HSZRJFAPSA-N
- Molecular Weight: 437.443
- Molecular Formula: C25H21F2NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,5-difluorophenyl)butanoic acid
- SMILES: O=C([C@H](NC(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CCC4=CC(F)=CC=C4F)O

Catalog No. 50-235-2007 Supplier STA Pharmaceutical Supplier No. STAAA1025110GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-hPhe(2,5-diF)-OH is a Amino Acid reagent (Subcategory: Homo-natural AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1260601-14-6
- MDL: MFCD07372306
- InChIKey: CMOCQWVOWRXWCO-HSZRJFAPSA-N
- Molecular Weight: 437.443
- Molecular Formula: C25H21F2NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,5-difluorophenyl)butanoic acid
- SMILES: O=C([C@H](NC(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CCC4=CC(F)=CC=C4F)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.