Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-3-Abz-Oh

Catalog No. 4795225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-952-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 47-952-25G Supplier Sigma Aldrich Supplier No. 479522.5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-3-Abz-Oh

Specifications

CAS 1282-37-7
Melting Point 178°C
Molecular Formula C10H10BF4Fe
Linear Formula C10H10BF4Fe
MDL Number MFCD00192174
Quantity 5 g
Synonym Bis(cyclopentadienyl)iron tetrafluoroborate; Dicyclopentadienyliron fluoborate; Dicyclopentadienyliron tetrafluoroborate
Molecular Weight (g/mol) 272.84 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.